missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ASK1 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF3575
This item is not returnable.
View return policy
Description
ASK1 Polyclonal specifically detects ASK1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| ASK1 | |
| Polyclonal | |
| Unconjugated | |
| Q99683 | |
| MAP3K5 | |
| E. coli-derived recombinant human ASK1 Lys1011-Asp1196 Accession # Q99683 | |
| 100 μg | |
| Primary | |
| Detects human ASK1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Apoptosis signal-regulating kinase 1, ASK-1, ASK1MEKK 5, EC 2.7.11, MAP/ERK kinase kinase 5, MAP3K5, MAPK/ERK kinase kinase 5, MAPKKK5EC 2.7.11.25, MEK kinase 5, MEKK5, MEKK5apoptosis signal regulating kinase 1, mitogen-activated protein kinase kinase kinase 5 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4217 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction