missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ATM Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB22901-SP
This item is not returnable.
View return policy
Description
ATM Monoclonal specifically detects ATM in Human samples. It is validated for Immunocytochemistry.
Specifications
| ATM | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| Q13315 | |
| ATM | |
| E. coli-derived recombinant human ATM Arg2138-Arg2400 Accession # Q13315 | |
| 25 μg | |
| Primary | |
| Detects human ATM in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human ATR is observed. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 664703 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated, ataxia telangiectasia mutated (includes complementation groups A, C and D), ATC, ATD, ATDC, ATE, ATM serine/threonine kinase, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1, TPLL | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 472 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction