missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AURKB (NM_004217, 2 a.a. - 344 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells

Product Code. 16017876
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16017876 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16017876 Supplier Abnova™ Supplier No. P4652.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Chromosomal segregation during mitosis as well as meiosis is regulated by kinases and phosphatases. The Aurora kinases associate with microtubules during chromosome movement and segregation. Aurora kinase B localizes to microtubules near kinetochores, specifically to the specialized microtubules called K-fibers, and Aurora kinase A (MIM 603072) localizes to centrosomes (Lampson et al., 2004 [PubMed 14767480]).[supplied by OMIM]

Specifications

Accession Number NM_004217
Concentration 0.196 ug/uL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 9212
Molecular Weight (g/mol) 70.073kDa
Name AURKB (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Purification Method GST Affinity Chromatography
Quality Control Testing 2 ug/lane SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing
Regulatory Status RUO
Gene Alias AIK2/AIM-1/AIM1/ARK2/AurB/IPL1/STK12/STK5
Common Name AURKB
Gene Symbol AURKB
Biological Activity 37pmol/μg x min
Species Baculovirus
Recombinant Recombinant
Protein Tag GST-His
Expression System Insect cell (Sf9) expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.