missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-H7/HHLA2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB80841R-025
This item is not returnable.
View return policy
Description
B7-H7/HHLA2 Monoclonal specifically detects B7-H7/HHLA2 in Human samples. It is validated for Flow Cytometry.
Specifications
| B7-H7/HHLA2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| B7H7, HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, HHLA2, Human endogenous retrovirus-H long terminal repeat-associating protein 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11148 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 907812 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Q9UM44 | |
| HHLA2 | |
| Human embryonic kidney cell line HEK293-derived human B7-H7/HHLA2 Met1-Asn344 Accession # Q9UM44 | |
| 25 Tests | |
| Primary | |
| Detects human B7-H7/HHLA2 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction