missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-H7/HHLA2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB80841-SP
This item is not returnable.
View return policy
Description
B7-H7/HHLA2 Monoclonal specifically detects B7-H7/HHLA2 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready.
Specifications
| B7-H7/HHLA2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B7H7, HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, HHLA2, Human endogenous retrovirus-H long terminal repeat-associating protein 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11148 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF | |
| 907812 | |
| Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 100-300 ng/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Q9UM44 | |
| HHLA2 | |
| Human embryonic kidney cell line HEK293-derived human B7-H7/HHLA2 Met1-Asn344 Accession # Q9UM44 | |
| 25 μg | |
| Primary | |
| Detects human B7-H7/HHLA2 in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction