missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BAFFR/TNFRSF13C Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB1162R-100UG
This item is not returnable.
View return policy
Description
BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Flow Cytometry.
Specifications
| BAFFR/TNFRSF13C | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human BAFF R/TNFRSF13C in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
| Flow Cytometry | |
| 2403C | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q96RJ3 | |
| Mouse myeloma cell line, NS0-derived human BAFF R/TNFRSF13C, Ser7-Ala71, Accession No. Q96RJ3 | |
| 100 μg | |
| 115650 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction