missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BAFFR/TNFRSF13C Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 12 publications
Brand: R&D Systems AF1162
This item is not returnable.
View return policy
Description
BAFFR/TNFRSF13C Polyclonal specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready.
Specifications
| BAFFR/TNFRSF13C | |
| Polyclonal | |
| Unconjugated | |
| Q96RJ3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 115650 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 3-15 ug/mL, Neutralization 3-15 ug/mL, CyTOF-ready | |
| TNFRSF13C | |
| Mouse myeloma cell line NS0-derived recombinant human BAFF R/TNFRSF13C Arg2-Ala71 Accession # Q96RJ3 | |
| 100 μg | |
| Primary | |
| Detects human BAFF R in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse BAFF R and less than 2% cross-reactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhNGF R, rhOPG, rhRANK, rhTNF RI, and rhTNF RII is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction