missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BAI3 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3965T-100UG
This item is not returnable.
View return policy
Description
BAI3 Monoclonal specifically detects BAI3 in Human samples. It is validated for Flow Cytometry.
Specifications
| BAI3 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| brain-specific angiogenesis inhibitor 3, dJ91B17.1 (brain-specific angiogenesis inhibitor 3), KIAA0550MGC133100 | |
| Mouse myeloma cell line NS0-derived recombinant human BAI3, Ala25-Thr880, Accession # O60242 | |
| 100 μg | |
| 577 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 409611 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| O60242 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human BAI3 in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction