missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BAMBI/NMA Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF921
This item is not returnable.
View return policy
Description
BAMBI/NMA Polyclonal specifically detects BAMBI/NMA in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| BAMBI/NMA | |
| Polyclonal | |
| Unconjugated | |
| Q13145 | |
| BAMBI | |
| Mouse myeloma cell line NS0-derived recombinant human BAMBI/NMA Glu27-Ala152 Accession # Q13145 | |
| 100 μg | |
| Primary | |
| Detects human BAMBI/NMA in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| BMP and activin membrane-bound inhibitor homolog, BMP and activin membrane-bound inhibitor homolog (Xenopus laevis), NMA, NMANon-metastatic gene A protein, Putative transmembrane protein NMA | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 25805 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu