missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BATF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8054-SP
This item is not returnable.
View return policy
Description
BATF Monoclonal specifically detects BATF in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| BATF | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q16520 | |
| BATF | |
| E. coli-derived recombinant human BATF Met1-Pro125 Accession # Q16520 | |
| 25 μg | |
| Primary | |
| Detects human BATF in ELISA and Western Blot. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 687706 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA2, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10538 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction