missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BCAM Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF148-SP
This item is not returnable.
View return policy
Description
BCAM/CD239 Polyclonal specifically detects BCAM/CD239 in Human samples. It is validated for Western Blot, Adhesion Blockade.
Specifications
| BCAM/CD239 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4059 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Adhesion Blockade | |
| LYOPH | |
| Western Blot 1 ug/mL, Adhesion Blockade | |
| CAA58449 | |
| BCAM | |
| Mouse myeloma cell line NS0-derived recombinant human BCAM Glu32-Ala547 Accession # CAA58449 | |
| 25 μg | |
| Primary | |
| Detects human BCAM in direct ELISAs and Western blots. In these formats, approximately 2% cross-reactivity with recombinant human (rh) ALCAM is observed and less than 1% cross-reactivity with rhPECAM, rhEpCAM, rhICAM-1, rhICAM-2, rhICAM-3, and rhVCAM-1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction