missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BCAR3 (aa 32-138) Control Fragment Recombinant Protein

Product Code. 30206097
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206097

Brand: Invitrogen™ RP90905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53244 (PA5-53244. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Breast tumors are initially dependent on estrogens for growth and progression and can be inhibited by anti-estrogens such as tamoxifen. However, breast cancers progress to become anti-estrogen resistant. Breast cancer anti-estrogen resistance gene 3 was identified in the search for genes involved in the development of estrogen resistance. The gene encodes a component of intracellular signal transduction that causes estrogen-independent proliferation in human breast cancer cells. The protein contains a putative src homology 2 (SH2) domain, a hall mark of cellular tyrosine kinase signaling molecules, and is partly homologous to the cell division cycle protein CDC48.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75815
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8412
Name Human BCAR3 (aa 32-138) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI131758; And34; AND-34; Bcar3; breast cancer anti-estrogen resistance 3; breast cancer antiestrogen resistance 3 protein; breast cancer anti-estrogen resistance protein 3; dJ1033H22.2 (breast cancer anti-estrogen resistance 3); KIAA0554; novel SH2-containing protein 2; NSP2; OTTHUMP00000011961; p130Cas-binding protein AND-34; SH2 domain-containing protein 3 B; SH2D3B; UNQ271/PRO308
Common Name BCAR3
Gene Symbol BCAR3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SPLAEHRPDAYQDVSIHGTLPRKKKGPPPIRSCDDFSHMGTLPHSKSPRQNSPVTQDGIQESPWQDRHGETFTFRDPHLLDPTVEYVKFSKERHIMDRTPEKLKKEL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.