missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Bcl-9 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB3996
This item is not returnable.
View return policy
Description
Bcl-9 Monoclonal specifically detects Bcl-9 in Human samples. It is validated for Western Blot.
Specifications
| Bcl-9 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| O00512 | |
| BCL9 | |
| E. coli-derived recombinant human Bcl-9 Met1009-Gly1328 Accession # O00512 | |
| 100 μg | |
| Primary | |
| Detects endogenous human Bcl-9 in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 417514 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B-cell CLL/lymphoma 9, B-cell CLL/lymphoma 9 protein, B-cell lymphoma 9 protein, bcl-9, Bcl9, BCL9L, LGS, MGC131591, Protein legless homolog | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 607 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction