missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BDNF APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC2481A
This item is not returnable.
View return policy
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| BDNF | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 627 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 35909 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| P23560 | |
| BDNF | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human BDNF His129-Arg247 Accession # P23560 | |
| 100 Tests | |
| Primary | |
| Detects human BDNF in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) NT-3, rhNT-4, or rh beta-NGF is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction