missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-2 Adrenergic R/ADRB2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB100401-100
This item is not returnable.
View return policy
Description
beta-2 Adrenergic R/ADRB2 Monoclonal specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| beta-2 Adrenergic R/ADRB2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ADRB2, ADRB2R, ADRBR, adrenergic, beta-2-, receptor, surface, B2AR, B2ARADRB2R, BAR, beta-2 adrenergic receptor, beta2 AdrenergicR, beta-2 AdrenergicR, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor | |
| NS0 mouse myeloma cell line transfected with human beta-2 Adrenergic R/ADRB2, Accession No. P07550 | |
| 100 μg | |
| 154.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 586107 | |
| Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| P07550 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human beta-2 Adrenergic R/ADRB2 in direct ELISAs. | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction