missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Bif1 (aa 84-162) Control Fragment Recombinant Protein

Product Code. 30197313
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197313

Brand: Invitrogen™ RP90706

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53340 (PA5-53340. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Bif1 gene encodes a protein similar to a protein in rodents which is induced by bone morphogenic protein 2 in vitro and may be involved in BMP2-induced transcription.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y371
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51100
Name Human Bif1 (aa 84-162) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA409932; AI314629; AU015566; Bax-interacting factor 1; BIF1; Bif-1; CGI-61; dJ612B15.2; EMBL:AAH79085.1}; endophilin b1; endophilin-B1; Kiaa0491; mKIAA0491; PPP1R70; protein phosphatase 1, regulatory subunit 70; SH3 domain containing GRB2 like endophilin B1; SH3 domain-containing GRB2-like protein B1; SH3-containing protein SH3GLB1; SH3-domain GRB2 like endophilin B1; SH3-domain GRB2-like B1 (endophilin); SH3-domain GRB2-like endophilin B1; SH3GLB1; sh3glb1 {ECO:0000312; testicular tissue protein Li 172
Common Name Bif1
Gene Symbol Sh3glb1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DRKAPSRINNPELLGQYMIDAGTEFGPGTAYGNALIKCGETQKRIGTADRELIQTSALNFLTPLRNFIEGDYKTIAKER
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.