Learn More
Human BLIMP1/PRDM1 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems IC36081R-025
Product Code. 18782837
  • € 309.00 / 25 tests
Description
BLIMP1/PRDM1 Monoclonal specifically detects BLIMP1/PRDM1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BLIMP1/PRDM1
Monoclonal
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Beta-interferon gene positive regulatory domain I-binding factor, beta-interferon gene positive-regulatory domain I binding factor, BLIMP-1, BLIMP1MGC118925, blmp-1, B-lymphocyte-induced maturation protein 1, MGC118922, MGC118923, Positive regulatory domain I-binding factor 1, PR domain containing 1, with ZNF domain, PR domain zinc finger protein 1, PR domain-containing protein 1, PRDI-BF1MGC118924, PRDI-binding factor 1, PRDI-binding factor-1, PRDM1, PR-domain zinc finger protein 1, ZNFPR1A1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
639
Human
Purified
Flow Cytometry
646702
Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
O75626
PRDM1
E. coli-derived recombinant human BLIMP1/PRDM1 Lys667-Cys789 Accession # O75626
25 Tests
Primary
Detects human BLIMP1/PRDM1 in Western blots.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.