missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMI-1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC33341R-100UG
This item is not returnable.
View return policy
Description
BMI-1 Monoclonal specifically detects BMI-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| BMI-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P35226 | |
| BMI1 | |
| E. coli-derived recombinant human BMI-1 Asp96-Gly326 Accession # P35226 | |
| RUO | |
| 648 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 384515 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| B lymphoma Mo-MLV insertion region 1 homolog, B lymphoma Mo-MLV insertion region 1 homolog (mouse), BMI1, BMI1 polycomb ring finger oncogene, FLVI2/BMI1, MGC12685, murine leukemia viral (bmi-1) oncogene homolog, PCGF4, polycomb complex protein BMI-1, polycomb group protein Bmi1, polycomb group ring finger 4, RING finger protein 51, RNF51, RNF51Polycomb group RING finger protein 4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human BMI-1 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction