missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMI-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB33341-SP
This item is not returnable.
View return policy
Description
BMI-1 Monoclonal specifically detects BMI-1 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Chromatin Immunoprecipitation (ChIP), CyTOF-ready.
Specifications
| BMI-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, CyTOF-ready | |
| P35226 | |
| BMI1 | |
| E. coli-derived recombinant human BMI-1 Asp96-Gly326 Accession # P35226 | |
| 25 μg | |
| Primary | |
| Detects human BMI-1 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| ChIP Assay, CyTOF, Flow Cytometry | |
| 384515 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B lymphoma Mo-MLV insertion region 1 homolog, B lymphoma Mo-MLV insertion region 1 homolog (mouse), BMI1, BMI1 polycomb ring finger oncogene, FLVI2/BMI1, MGC12685, murine leukemia viral (bmi-1) oncogene homolog, PCGF4, polycomb complex protein BMI-1, polycomb group protein Bmi1, polycomb group ring finger 4, RING finger protein 51, RNF51, RNF51Polycomb group RING finger protein 4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 648 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction