missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BMP-15 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
BMP15 Synthetic Peptide, PEP-1443, from Invitrogen.
BMP15, or GDF9B is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily that are involved in embryonic development and adult tissue homeostasis. BMP15 is expressed exclusively in the oocyte. It is an oocyte-specific growth/differentiation factor that may be involved in oocyte maturation and follicular development. Defects in BMP15 are the cause of ovarian dysgenesis type 2. BMP15 could be used as an oogenesis marker to track human amniotic fluid stem cells differentiation into the oocyte-like cells.
Specifications
Specifications
| Gene ID (Entrez) | 9210 |
| Species | Human |
| Purification Method | purified |
| Accession Number | O95972 |
| Common Name | Human BMP-15 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction