missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BMP-9 ELISA Kit

Product Code. 17155279
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17155279 5 x 96 Tests Each
17114803 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17155279 Supplier Invitrogen™ Supplier No. EH49RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human BMP-9 quantitates human BMP-9 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human BMP-9.

The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in rodents suggest that this protein plays a role in the adult liver and in differentiation of cholinergic central nervous system neurons.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number BMP; BMP9; BMP-9; Bone morphogenetic protein; Bone morphogenetic protein 9; Gdf2; GDF-2; growth differentiation factor 2; growth/differentiation factor 2; HHT5
Assay Range 32 to 8,000 pg/mL
Assay Sensitivity 32 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2658
Gene Symbol GDF2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target BMP9
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.