missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BPI Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC74681U-100UG
This item is not returnable.
View return policy
Description
BPI Monoclonal specifically detects BPI in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| BPI | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| bactericidal permeability-increasing protein, bactericidal/permeability-increasing protein, BPIFD1, CAP 57, rBPI | |
| Human embryonic kidney cell line HEK-293-derived transfected with human BPI, Val32-Lys487, Accession # P17213 | |
| 100 μg | |
| 671 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 971526 | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| P17213 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human BPI in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction