missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Bradykinin RB2/BDKRB2 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9434S-100UG
This item is not returnable.
View return policy
Description
Bradykinin RB2/BDKRB2 Monoclonal specifically detects Bradykinin RB2/BDKRB2 in Human samples. It is validated for Flow Cytometry.
Specifications
| Bradykinin RB2/BDKRB2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
| B2 bradykinin receptor, B2R, BDKRB2, BK2, BK-2, BK-2 receptor, BKR2, bradykinin receptor B2, BradykininRB2, BRB2, DKFZp686O088 | |
| NS0 mouse myeloma cell line transfected with human RB2/BDKRB2, Met1-Gln391, Accession No. P30411 | |
| 100 μg | |
| 624.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 471902 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P30411 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human RB2/BDKRB2 in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction