missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BRCA1 N-Terminus Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2210-SP
This item is not returnable.
View return policy
Description
BRCA1 Polyclonal specifically detects BRCA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| BRCA1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 672 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 3 ug/500 ug cell lysate | |
| P38398 | |
| BRCA1 | |
| E. coli-derived recombinant human BRCA1 N-Terminus Met1-Gly200 Accession # P38398 | |
| 25 μg | |
| Primary | |
| Detects human BRCA1 N-Terminus in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction