missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BRD1 (aa 907-1053) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-81916 (PA5-81916. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types.
Specifications
Specifications
| Accession Number | O95696 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 23774 |
| Name | Human BRD1 (aa 907-1053) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1110059H06Rik; AI316859; BR140-like protein; BRD1; BRL; Bromodomain and PHD finger-containing protein 2; bromodomain containing 1; bromodomain-containing protein 1; BRPF1; BRPF2; DKFZp686F0325; mKIAA4191; RP3-522J7.4 |
| Common Name | BRD1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction