missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BTN3A1/2/3 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7136A-025
This item is not returnable.
View return policy
Description
BTN3A1/2/3 Monoclonal specifically detects BTN3A1/2/3 in Human samples. It is validated for Flow Cytometry.
Specifications
| BTN3A1/2/3 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| BT3.1, BTF5, BTN3A1, butyrophilin, subfamily 3, member A1, CD277, dJ45P21.3 (butyrophilin, subfamily 3, member A1) | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 11119 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 849203 | |
| Flow Cytometry 10 uL/10^6 cells | |
| O00481 | |
| BTN3A1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD277/BTN3A1, Met1-Gly254, Accession # O00481 | |
| 25 Tests | |
| Primary | |
| Detects human BTN3A1, BTN3A2 and BTN3A3 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction