missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BTNL8 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB9359G-100UG
This item is not returnable.
View return policy
Description
BTNL8 Monoclonal specifically detects BTNL8 in Human samples. It is validated for Flow Cytometry.
Specifications
| BTNL8 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| BTN9.2, butyrophilin-like 8, butyrophilin-like protein 8, UNQ702/PRO1347 | |
| Human embryonic kidney cell line HEK-293-derived Human BTNL8 Gln18-Lys238 Accession # Q6UX41-1 | |
| 100 μg | |
| 79908 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 2187b | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q6UX41-1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human BTNL8 in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction