Learn More
Human BTNL8 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB93591-100
 View more versions of this product
Product Code. 18171751
  • € 449.00 / 100µg
Description
BTNL8 Monoclonal antibody specifically detects BTNL8 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BTNL8
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
BTN9.2, butyrophilin-like 8, butyrophilin-like protein 8, UNQ702/PRO1347
Human embryonic kidney cell HEK293-derived human BTNL8 protein, Gln18-Lys238, Accession # Q6UX41-1
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
IgG1
Immunohistochemistry
1047350
Immunohistochemistry 5-25 ug/mL
Q6UX41-1
Mouse
Protein A or G purified from cell culture supernatant
RUO
79908