Learn More
Human C-Reactive Protein/CRP Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB17071-100
 View more versions of this product
Product Code. 18762808
  • € 530.00 / 100µg
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C-Reactive Protein/CRP
Monoclonal
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P02741
CRP
Human plasma-derived CRP Accession # P02741
100 μg
Primary
Detects human CRP in direct ELISAs.
Human
Supernatant
ELISA
232007
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
1401
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b