missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human C1 Inhibitor Recombinant Protein, PeproTech®

Product Code. p-7246172
Click to view available options
Quantity:
1 mg
200 μg
50 μg
500 μg
Unit Size:
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17891553

Brand: Gibco™ 130201MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human C1 Inhibitor is a highly glycosylated glycoprotein containing 478 amino acid residues (52.8kDa), corresponding to amino acids 23-500 of the C1 inhibitor precursor, and is fully functional in its ability to inhibit the C1 complex. Glycosylated C1 Inhibitor migrates at an apparent molecular weight of approximately 80-90 kDa by SDS PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

C1-inhibitor (C1INH) functions as a regulator of the activation of the classical complement pathway and of the contact activation system of kinin generation and coagulation. Its primary biologically relevant target proteinases are C1r, C1s, coagulation factors XIa and XIIa, and plasma kallikrein. C1-inhibitor consists of one polypeptide chain and has a molecular mass of ∽104 kDa. Serum concentration has been determined to 180 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05155
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 710
Molecular Weight (g/mol) 52.8 kDa
Name Human C1 Inhibitor
Quantity 1 mg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C1 esterase inhibitor; C1 Inh; C1 inhibitor; C1IN; C1Inh; C1-inhibiting factor; C1nh; complement component 1 inhibitor; complement component 1 inhibitor (angioedema, hereditary); esterase inhibitor; factor XIIa inhibitor; HAE1; HAE2; IC1; Plasma protease C1 inhibitor; serine (or cysteine) peptidase inhibitor, clade G, member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serine (or cysteine) proteinase inhibitor, clade G, member 1; serine/cysteine proteinase inhibitor clade G member 1; serpin family G member 1; serpin G1; serpin peptidase inhibitor clade G member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serpin peptidase inhibitor, clade G, member 1; SERPING1; XIIaINH
Common Name C1 inhibitor
Gene Symbol SERPING1
Biological Activity Measured by its ability to inhibit recombinant human complement component C1a cleavage of a colorimetric peptide substrate, N Carbobenzyloxy-Lys-ThioBenzyl ester (Z-K-SBzl). The expected IC50 is ≤ 2.6 nM.
Conjugate Unconjugated
Recombinant Recombinant
Sequence NPNATSSSSQ DPESLQDRGE GKVATTVISK MLFVEPILEV SSLPTTNSTT NSATKITANT TDEPTTQPTT EPTTQPTIQP TQPTTQLPTD SPTQPTTGSF CPGPVTLCSD LESHSTEAVL GDALVDFSLK LYHAFSAMKK VETNMAFSPF SIASLLTQVL LGAGENTKTN LESILSYPKD FTCVHQALKG FTTKGVTSVS QIFHSPDLAI RDTFVNASRT LYSSSPRVLS NNSDANLELI NTWVAKNTNN KISRLLDSLP SDTRLVLLNA IYLSAKWKTT FDPKKTRMEP FHFKNSVIKV PMMNSKKYPV AHFIDQTLKA KVGQLQLSHN LSLVILVPQN LKHRLEDMEQ ALSPSVFKAI MEKLEMSKFQ PTLLTLPRIK VTTSQDMLSI MEKLEFFDFS YDLNLCGLTE DPDLQVSAMQ HQTVLELTET GVEAAAASAI SVARTLLVFE VQQPFLFVLW DQQHKFPVFM GRVYDPRA
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.