missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human C10orf99 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC104161V-100UG
This item is not returnable.
View return policy
Description
GPR15L Monoclonal antibody specifically detects GPR15L in Human samples. It is validated for Flow Cytometry
Specifications
| GPR15L | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azid | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 387695 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 1020904 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/106 cells | |
| AP57, AP-57, C10orf99 chromosome 10 open reading frame 99, GPR15L, UNQ1833 | |
| E. coli-derived human C10orf99 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction