missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human C1qTNF5 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC3167N-100UG
This item is not returnable.
View return policy
Description
C1qTNF5/CTRP5 Monoclonal specifically detects C1qTNF5/CTRP5 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| C1qTNF5/CTRP5 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| C1q and tumor necrosis factor related protein 5, C1qTNF5, Complement C1q Tumor Necrosis Factor-Related Protein 5 Precursor Variant 3, CTRP5LORDDKFZp586B0621, LORD 6, Myonectin 3 | |
| Mouse myeloma cell line NS0-derived recombinant human C1qTNF5 Ser16-Ala243 (Gln44Arg) Accession # Q9BXJ0 | |
| RUO | |
| 114902 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 332923 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9BXJ0 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human C1qTNF5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) C1qTNF1 or rhC1qTNF4 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction