missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CA125/MUC16 Alexa Fluor 700-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB5609N-100UG
This item is not returnable.
View return policy
Description
CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for Flow Cytometry.
Specifications
| CA125/MUC16 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, MUC16, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CA125/MUC-16, Met13360-Gln14347 (Met13472Thr & Gln13957Lys), Accession # NP_078966.2 | |
| 100 μg | |
| 94025 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 986808 | |
| Flow Cytometry 0.25-1μg/106 cells | |
| NP_078966.2 | |
| Mouse | |
| Protein A or G purified from ascites | |
| Primary | |
| Detects human CA125/MUC-16 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction