missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cadherin-11 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB17901R-100UG
This item is not returnable.
View return policy
Description
Cadherin-11 Monoclonal specifically detects Cadherin-11 in Human samples. It is validated for Flow Cytometry.
Specifications
| Cadherin-11 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAA35622 | |
| CDH11 | |
| Mouse myeloma cell line NS0-derived recombinant human Cadherin-11 Phe23-Thr617 Accession # AAA35622 | |
| RUO | |
| 1009 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 667039 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CAD11OSF-4, cadherin 11, type 2, OB-cadherin (osteoblast), cadherin-11, Cadherin11, CDH11, CDHOB, OB, OB-cadherin, Osteoblast cadherin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Cadherin-11 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Cadherin-4, -6, -8, -12, -13, -17, rhE-Cadherin, rhN-Cadherin, rhP-Cadherin, or rhVE-Cadherin is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction