missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Calreticulin-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2927-SP
This item is not returnable.
View return policy
Description
Calreticulin-2/CALR3 Monoclonal specifically detects Calreticulin-2/CALR3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Calreticulin-2/CALR3 | |
| Monoclonal | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q96L12 | |
| CALR3 | |
| E. coli-derived recombinant human Calreticulin-2 Thr20-Leu384 Accession # Q96L12 | |
| 25 μg | |
| Primary | |
| Detects human Calreticulin-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Calreticulin is observed. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 321007 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CALR3, calreticulin 2, calreticulin 3, Calreticulin2, Calreticulin-2, calreticulin-3, cancer/testis antigen 93, Crt2, CRT2calreticulin-2, CT93, FLJ25355, MGC26577 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 125972 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction