missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cannabinoid R1/CB1/CNR1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3834R-100UG
This item is not returnable.
View return policy
Description
Cannabinoid R1/CB1/CNR1 Monoclonal specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Cannabinoid R1/CB1/CNR1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_057167 | |
| CNR1 | |
| HEK293 human embryonic kidney cell line transfected with human Cannabinoid R1/CB1/CNR1 Met1-Leu472 Accession # NP_057167 | |
| RUO | |
| 1268 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 368302 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CANN6, CANN6CB1R, cannabinoid receptor 1 (brain), CannabinoidR1, CB1, CB1A, CB1K5, CB1R, CB-R, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR, CNR1, SKR6R | |
| Mouse | |
| 100 μg | |
| Primary | |
| Stains Cannabinoid R1/CB1/CNR1-transfected cells but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction