missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Carbonic Anhydrase IX/CA9 Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody
€ 218.00 - € 536.00
Specifications
| Antigen | Carbonic Anhydrase IX/CA9 |
|---|---|
| Clone | 1349A |
| Dilution | Immunohistochemistry 1-3 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30018986
![]() |
Novus Biologicals™
MAB11526-SP |
25 μg |
€ 218.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019009
|
Novus Biologicals™
MAB11526-100 |
100 μg |
€ 536.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbonic Anhydrase IX/CA9 Monoclonal antibody specifically detects Carbonic Anhydrase IX/CA9 in Human samples. It is validated for ImmunohistochemistrySpecifications
| Carbonic Anhydrase IX/CA9 | |
| Immunohistochemistry 1-3 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CA9, CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
| Mouse myeloma cell line, NS0-derived human Carbonic Anhydrase IX/CA9, Pro59-Asp414, Accession # Q16790 | |
| Primary | |
| Reconstitute lyophilized material at 0.2mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1349A | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q16790 | |
| 768 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts