missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carbonic Anhydrase IX/CA9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB21881-SP
This item is not returnable.
View return policy
Description
Carbonic Anhydrase IX/CA9 Monoclonal specifically detects Carbonic Anhydrase IX/CA9 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Carbonic Anhydrase IX/CA9 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q16790 | |
| CA9 | |
| Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase IX/CA9 Pro59-Asp414 Accession # Q16790 | |
| 25 μg | |
| Primary | |
| Detects human Carbonic Anhydrase IX/CA9 in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 303104 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CA9, CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 768 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction