missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carbonic Anhydrase VI Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC29391T-100UG
This item is not returnable.
View return policy
Description
Carbonic Anhydrase VI Monoclonal specifically detects Carbonic Anhydrase VI in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Carbonic Anhydrase VI | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| EAW71606 | |
| CA6 | |
| Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase VI Gln18-Asn308 Accession # EAW71606 | |
| RUO | |
| 765 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 401809 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CA6, Carbonate dehydratase VI, carbonic anhydrase 6, carbonic anhydrase VIGUSTIN, CA-VI, EC 4.2.1.1, Gustin, MGC21256, Salivary carbonic anhydrase, Secreted carbonic anhydrase | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Carbonic Anhydrase VI in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Carbonic Anhydrase 1, 2, 3, 4, 5A, 5B, 7, 8, or 9 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction