Learn More
Invitrogen™ Human Carbonic Anhydrase XII (aa 84-144) Control Fragment Recombinant Protein
Product Code. 30198077
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30198077
Brand: Invitrogen™ RP107912
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84921 (PA5-84921. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Carbonic Anhydrase XII is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number O43570
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 771
Name Human Carbonic Anhydrase XII (aa 84-144) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310047E01Rik; AI314958; CA12; Car12; Carbonate dehydratase XII; carbonic anhydrase 12; Carbonic anhydrase XII; carbonic anyhydrase 12; carbonic dehydratase; CAXII; CA-XII; HsT18816; membrane-bound carbonic anhydrase 12; T18816; tumor antigen HOM-RCC-3.1.3
Common Name Carbonic Anhydrase XII
Gene Symbol CA12
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NKQFLLTNNGHSVKLNLPSDMHIQGLQSRYSATQLHLHWGNPNDPHGSEHTVSGQHFAAEL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining