Learn More
Invitrogen™ Human Carbonic Anhydrase XIII (aa 10-92) Control Fragment Recombinant Protein
Product Code. 30203696
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30203696
Brand: Invitrogen™ RP93943
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55154 (PA5-55154. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Carbonic anhydrases (CAs) are a family of zinc metalloenzymes. For background information on the CA family, see MIM 114800.[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q8N1Q1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 377677
Name Human Carbonic Anhydrase XIII (aa 10-92) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310075C21Rik; Ca13; Car13; Carbonate dehydratase XIII; carbonic anhydrase 13; Carbonic anhydrase XIII; CAXIII; CA-XIII; RGD1560453
Common Name Carbonic Anhydrase XIII
Gene Symbol Ca13
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EHNGPIHWKEFFPIADGDQQSPIEIKTKEVKYDSSLRPLSIKYDPSSAKIISNSGHSFNVDFDDTENKSVLRGGPLTGSYRLR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining