missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carboxylesterase 1/CES1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB4920
This item is not returnable.
View return policy
Description
Carboxylesterase 1/CES1 Monoclonal specifically detects Carboxylesterase 1/CES1 in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.
Specifications
| Carboxylesterase 1/CES1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation 25 ug/mL | |
| P23141 | |
| CES1 | |
| Mouse myeloma cell line NS0-derived recombinant human Carboxylesterase1/CES-1 His19-Glu563 Accession # P23141 | |
| 100 μg | |
| Primary | |
| Detects human Carboxylesterase1/CES-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse (rm) CES-3 and no cross-reactivity with rmCES-2, rmCES-5, or recombinant human CES-2 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 544623 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACAT, acyl coenzyme A:cholesterol acyltransferase, Acyl-coenzyme A:cholesterol acyltransferase, Brain carboxylesterase hBr1, carboxylesterase 1, carboxylesterase 1 (monocyte/macrophage serine esterase 1), carboxylesterase 2 (liver), CEH, Ces-1, CES1, CES1A2, CES2EC 3.1.1.1, cholesteryl ester hydrolase, Cocaine carboxylesterase, EC 3.1.1, egasyn, HMSE1, HMSECES1A1, human monocyte/macrophage serine esterase 1, liver carboxylesterase 1, MGC117365, Monocyte/macrophage serine esterase, PCE-1, REH, Retinyl ester hydrolase, Serine esterase 1, Ses-1, SES1, SES1TGH, TGH, Triacylglycerol hydrolase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1066 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction