missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CARF Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4195
This item is not returnable.
View return policy
Description
CARF/CDKN2AIP Polyclonal specifically detects CARF/CDKN2AIP in Human samples. It is validated for Western Blot.
Specifications
| CARF/CDKN2AIP | |
| Polyclonal | |
| Unconjugated | |
| Q9NXV6 | |
| CDKN2AIP | |
| E. coli-derived recombinant human CARF Ser379-Leu579 Accession # Q9NXV6 | |
| 100 μg | |
| Primary | |
| Detects human CARF in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| CARFCDKN2A-interacting protein, CDKN2A interacting protein, CDKN2AIP, collaborates/cooperates with ARF (alternate reading frame) protein, Collaborator of ARF, FLJ20036 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 55602 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction