missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carnosine Dipeptidase 1/CNDP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2489
This item is not returnable.
View return policy
Description
Carnosine Dipeptidase 1/CNDP1 Monoclonal specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.
Specifications
| Carnosine Dipeptidase 1/CNDP1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q96KN2 | |
| CNDP1 | |
| Mouse myeloma cell line NS0-derived recombinant human Carnosine Dipeptidase 1/CNDP1 Pro28-His507 Accession # Q96KN2 | |
| 100 μg | |
| Primary | |
| Detects human Carnosine Dipeptidase 1/CNDP1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CNDP2 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| 352216 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CNDP1, CPGL2, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84735 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction