missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human Caspase-9 Platinum ELISA Kit

For the quantitative detection of human Caspase-9.

Merk:  Invitrogen™ BMS2025TEN

Overige gegevens : Gewicht : 0.23750kg

 Bekijk meerdere types van dit product via de

Produktcode. 15541517

  • € 3055.00 / 10 stuks


Geschatte verzenddatum
bij leverancier 22-02-2019
Toevoegen aan winkelmandje

Inclusief: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Caspase-9
Anti-human Caspase-9 polyclonal Detection Antibody (rabbit)
Anti-rabbit-IgG-HRP
Human Caspase-9 Standard lyophilized, 200ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Lysis Buffer 10x
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

Beschrijving

Beschrijving

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specificatie

Specificatie

P55211
HRP
842
Pre-coated 96 well plate, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Detection Antibody, anti-rabbit HRP conjugate, Lysis Buffer, Wash Buffer, Chromogen, Stop Solution
10 x 96 tests
0.4ng/mL
50μL
Human
1 hr 20 min
Caspase 9
Apoptotic protease MCH-6, APAF-3, ICE-LAP6
6.6%
1.6-100 ng/mL
ELISA
Colorimetric Microplate Reader
ELISA
RUO
Cell culture supernatant, cell lysate, serum
Cell Lysates (50μL), Serum (50μL), Supernatants (50μL)
2-8°C
3 hr 10 min
CASP9
9%
HRP
Video's

For Research Use Only