missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin B Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF953
This item is not returnable.
View return policy
Description
Cathepsin B Polyclonal specifically detects Cathepsin B in Human samples. It is validated for Western Blot, Immunohistochemistry, ELISA Detection (Matched Antibody Pair).
Specifications
| Cathepsin B | |
| Polyclonal | |
| Biotin | |
| P07858 | |
| CTSB | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin B Arg18-Ile339 Accession # P07858 | |
| 50 μg | |
| Primary | |
| Detects human Cathepsin B in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) Cathepsin A, C, D, E, L, S, V, X/Z/P, rhCystatin C, D, SA, recombinant mouse (rm) Cystatin C, D, SA, and rmCathepsin B is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| APP secretase, APPS, cathepsin B, Cathepsin B1, CPSBamyloid precursor protein secretase, CTSB, cysteine protease, EC 3.4.22, EC 3.4.22.1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1508 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction