missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin D Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF1014
This item is not returnable.
View return policy
Description
Cathepsin D Polyclonal specifically detects Cathepsin D in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Cathepsin D | |
| Polyclonal | |
| Biotin | |
| P07339 | |
| CTSD | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin D Leu21-Leu412 Accession # P07339 | |
| 50 μg | |
| Primary | |
| Detects human Cathepsin D in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse Cathepsin D is observed, and less than 1% cross-reactivity with recombinant human (rh) Cathepsin A, rhCathepsin B, rhCathepsin C, rhCathepsin L, and rhCathepsin X/Z/P is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, CTSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1509 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction