missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin G, Synthetic Peptide, Invitrogen™
Synthetic Peptide
Brand: Invitrogen GSC00003P
Additional Details : Weight : 0.01000kg
Description
Human Cathepsin G Synthetic Peptide, GSC00003P, from Invitrogen.
GSC00003P is the immunizing peptide for the rabbit antibody to human Cathepsin G.GSC00003P may be used as a control in various immunology applications.Maintain lyophilized product refrigerated at 4ºC for up to 12 months. After reconstitution, keep at -20ºC.Reconstitute with PBS or distilled water.Specifications
1511 | |
purified | |
Human Cathepsin G | |
Synthetic Peptide | |
peptide with no preservative | |
CTSG protein, Cathepsin G, MGC23078 | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P08311 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized | |
Control | |
CTSG | |
No preservative | |
50 μg |