missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin H Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7516
This item is not returnable.
View return policy
Description
Cathepsin H Polyclonal specifically detects Cathepsin H in Human samples. It is validated for Western Blot.
Specifications
| Cathepsin H | |
| Polyclonal | |
| Unconjugated | |
| CAA34734 | |
| CTSH | |
| Chinese hamster ovary cell line CHO-derived recombinant human Cathepsin H Ala23-Val335 Accession # CAA34734 | |
| 100 μg | |
| Primary | |
| Detects human Cathepsin H in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse Cathepsin H is observed, and less than 1% cross-reactivity with recombinant human Cathepsin L2 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoassay | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Knockout Validated | |
| ACC-4, ACC-5, aleurain, cathepsin B3, cathepsin BA, cathepsin H, CPSB, CTSH, DKFZp686B24257, EC 3.4.22, EC 3.4.22.16, MGC1519, minichain, N-benzoylarginine-beta-naphthylamide hydrolase | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1512 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction