missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCDC134 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7784-SP
This item is not returnable.
View return policy
Description
CCDC134 Monoclonal specifically detects CCDC134 in Human samples. It is validated for Western Blot.
Specifications
| CCDC134 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9H6E4 | |
| CCDC134 | |
| Mouse myeloma cell line NS0-derived recombinant human CCDC134 Thr23-Leu229 Accession # Q9H6E4 | |
| 25 μg | |
| Primary | |
| Detects human CCDC134 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant mouse CCDC134 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 814208 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| coiled-coil domain containing 134, coiled-coil domain-containing protein 134, dJ821D11.3, FLJ22349, MGC21013 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 79879 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction